Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-boc-peg4-bromide | 1076199-21-7 | MFCD09840081 | 98.0% | 356.25 g/mol | C13H26BrNO5 | 500 MG
SDP

Catalog No. 5000204115
Encompass_Preferred

N-boc-peg4-bromide is a Boc-protected PEG4 bromide linker used as a PEG spacer and a reactive alkyl bromide for conjugation chemistry. It is intended for research use in small-molecule conjugation workflows, including PROTAC and cleavable linker synthesis, and has specified solubility and storage recommendations.

  • Boc-protected amine with terminal bromide for alkylation.
  • Designed as a PEG4 spacer to increase solubility and flexibility.
  • Soluble in DMSO; recommended concentration up to 100 mg/mL with ultrasonic assistance.
  • Purity ≥98.0% as supplied for research applications.
  • Available in multiple pack sizes, including a 500 mg pack.

Catalog No. 50-002-04115 Supplier Medchemexpress LLC Supplier No. HYW046471500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-boc-peg4-bromide is a Boc-protected PEG4 bromide linker used as a PEG spacer and a reactive alkyl bromide for conjugation chemistry. It is intended for research use in small-molecule conjugation workflows, including PROTAC and cleavable linker synthesis, and has specified solubility and storage recommendations.

  • Boc-protected amine with terminal bromide for alkylation.
  • Designed as a PEG4 spacer to increase solubility and flexibility.
  • Soluble in DMSO; recommended concentration up to 100 mg/mL with ultrasonic assistance.
  • Purity ≥98.0% as supplied for research applications.
  • Available in multiple pack sizes, including a 500 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.