Learn More
Medchemexpress LLC HY-W046471 250mg Medchemexpress, N-Boc-PEG4-bromide CAS:1076199-21-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW046471250MG
Encompass_Preferred
Medchemexpress, HY-W046471 250mg N-Boc-PEG4-bromide CAS:1076199-21-7 N-Boc-PEG4-bromide is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3484