Learn More
Medchemexpress LLC N-Boc-PEG5-bromide | 1392499-32-9 | MFCD31536756 | 98.0% | 400.31 g/mol | C15H30BrNO6 | 1 G
SDP
Catalog No. 5000202767
Encompass_Preferred
N-Boc-PEG5-bromide is a Boc-protected polyethylene glycol (PEG5) bromide linker reagent used in organic synthesis for assembling antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) linkers. It supplies a protected amine and a reactive terminal bromide for nucleophilic substitution and coupling reactions, and is provided at high purity for research use.
  • Provides a Boc-protected amine for temporary protection during synthesis.
  • Offers a terminal bromide suitable for nucleophilic substitution and coupling reactions.
  • PEG5 spacer increases solubility and flexibility in linker constructs.
  • High purity appropriate for research and linker assembly.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-02767 Supplier Medchemexpress LLC Supplier No. HY1207021G
N-Boc-PEG5-bromide is a Boc-protected polyethylene glycol (PEG5) bromide linker reagent used in organic synthesis for assembling antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) linkers. It supplies a protected amine and a reactive terminal bromide for nucleophilic substitution and coupling reactions, and is provided at high purity for research use.
  • Provides a Boc-protected amine for temporary protection during synthesis.
  • Offers a terminal bromide suitable for nucleophilic substitution and coupling reactions.
  • PEG5 spacer increases solubility and flexibility in linker constructs.
  • High purity appropriate for research and linker assembly.
  • Stable under recommended storage conditions for extended shelf life.