Learn More
Medchemexpress LLC N-Boc-PEG5-bromide | 1392499-32-9 | MFCD31536756 | 98.0% | 400.31 g·mol⁻¹ | C15H30BrNO6 | 100 MG
SDP
Catalog No. 5000204042
Encompass_Preferred
N-Boc-PEG5-bromide is a Boc-protected polyethylene glycol (PEG) linker bearing a terminal bromide functional group. It is used as a reactive building block in the synthesis of antibody-drug conjugates (ADCs) and PROTACs to introduce a five-unit PEG spacer that improves solubility and provides a reactive handle for conjugation chemistry.
  • Boc-protected amine allows selective deprotection under acidic conditions.
  • Terminal bromide enables alkylation and nucleophilic substitution reactions.
  • Five-unit PEG spacer improves aqueous solubility and linker flexibility.
  • High purity suitable for research applications (98.0%).
  • Supplied as a solid and available in small-scale pack sizes including 100 MG.
Catalog No. 50-002-04042 Supplier Medchemexpress LLC Supplier No. HY120702100MG
N-Boc-PEG5-bromide is a Boc-protected polyethylene glycol (PEG) linker bearing a terminal bromide functional group. It is used as a reactive building block in the synthesis of antibody-drug conjugates (ADCs) and PROTACs to introduce a five-unit PEG spacer that improves solubility and provides a reactive handle for conjugation chemistry.
  • Boc-protected amine allows selective deprotection under acidic conditions.
  • Terminal bromide enables alkylation and nucleophilic substitution reactions.
  • Five-unit PEG spacer improves aqueous solubility and linker flexibility.
  • High purity suitable for research applications (98.0%).
  • Supplied as a solid and available in small-scale pack sizes including 100 MG.