Learn More
Apexbio Technology LLC N-BOC-PEG5-BROMIDE-5G
SDP
Catalog No. 5000569121
Encompass_Preferred
N-Boc-PEG5-bromide (CAS No 1392499-32-9) is a compound utilized in biomedical research particularly within the fields of chemical biology and drug development Originating from synthetic chemistry it acts as a bromide-functionalized polyethylene glycol (PEG) derivative offering a hydrophilic linker that facilitates the delivery of bioactive molecules The compound is employed in modification of proteins and peptides enhancing solubility and stability In terms of in vitro activity it operates in nanomolar to low micromolar concentrations modulating specific cellular pathways by serving as a molecular probe Its application is extensive in cell models and animal studies aiding in drug screening processes to evaluate efficacy and mechanism of new therapeutics Experimental concentrations typically depend on the specific research design allowing for versatile usage tailored to particular experimental setups
Catalog No. 50-005-69121 Supplier Apexbio Technology LLC Supplier No. BA24255000
N-Boc-PEG5-bromide (CAS No 1392499-32-9) is a compound utilized in biomedical research particularly within the fields of chemical biology and drug development Originating from synthetic chemistry it acts as a bromide-functionalized polyethylene glycol (PEG) derivative offering a hydrophilic linker that facilitates the delivery of bioactive molecules The compound is employed in modification of proteins and peptides enhancing solubility and stability In terms of in vitro activity it operates in nanomolar to low micromolar concentrations modulating specific cellular pathways by serving as a molecular probe Its application is extensive in cell models and animal studies aiding in drug screening processes to evaluate efficacy and mechanism of new therapeutics Experimental concentrations typically depend on the specific research design allowing for versatile usage tailored to particular experimental setups