Learn More
Medchemexpress LLC HY-P1795 1mg Medchemexpress, N-Boc-Phe-Leu-Phe-Leu-Phe CAS:148182-34-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP17951MG
Encompass_Preferred
Medchemexpress, HY-P1795 1mg N-Boc-Phe-Leu-Phe-Leu-Phe CAS:148182-34-7 N-Boc-Phe-Leu-Phe-Leu-Phe (Boc-FLFLF) is a formyl peptide receptor 1 (FPR1) antagonist, which increases pain effects and inhibits antinociceptive activity of annexin [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4245