Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-phenylalanyl glycine | 25616-33-5 | MFCD00134859 | >97.0% | 322.36 g/mol | C16H22N2O5 | 5 G
SDP

Catalog No. 5000210112
Encompass_Preferred

Boc-Phe-Gly-OH is an N-Boc-protected phenylalanyl glycine derivative supplied as a white to off-white solid for research and synthetic applications. It is commonly used as a building block in peptide synthesis and as a precursor in small-molecule synthesis where a Boc-protected phenylalanyl-glycine moiety is required.

  • Molecular formula: C16H22N2O5
  • Molecular weight: 322.36 g/mol
  • Purity: >97.0%
  • Physical form: solid, white to off-white powder
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years)
  • Packaging: available in multiple sizes including 5 g

Catalog No. 50-002-10112 Supplier Medchemexpress LLC Supplier No. HYW0488295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Phe-Gly-OH is an N-Boc-protected phenylalanyl glycine derivative supplied as a white to off-white solid for research and synthetic applications. It is commonly used as a building block in peptide synthesis and as a precursor in small-molecule synthesis where a Boc-protected phenylalanyl-glycine moiety is required.

  • Molecular formula: C16H22N2O5
  • Molecular weight: 322.36 g/mol
  • Purity: >97.0%
  • Physical form: solid, white to off-white powder
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years)
  • Packaging: available in multiple sizes including 5 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.