Learn More
Medchemexpress LLC N-Boc-piperazine | 57260-71-6 | 99.99% | 186.25 | 25 G
SDP
Catalog No. 5000402869
Encompass_Preferred
N-Boc-piperazine is an alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTAC PD-1/PD-L1 degrader-1. This product is for research use only. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTAC PD-1/PD-L1 degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-02869 Supplier Medchemexpress LLC Supplier No. 25GHY30105
N-Boc-piperazine is an alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTAC PD-1/PD-L1 degrader-1. This product is for research use only. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTAC PD-1/PD-L1 degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only