Learn More
Medchemexpress LLC N-Boc-piperazine | 57260-71-6 | 99.99% | 186.25 | 50 G
SDP
Catalog No. 5000404814
Encompass_Preferred
N-Boc-piperazine is an Alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTAC PD-1/PD-L1 degrader-1. This product is for research use only and not sold to patients.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTAC PD-1/PD-L1 degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-04814 Supplier Medchemexpress LLC Supplier No. HY3010550G
N-Boc-piperazine is an Alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTAC PD-1/PD-L1 degrader-1. This product is for research use only and not sold to patients.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTAC PD-1/PD-L1 degrader-1
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins