Learn More
Medchemexpress LLC N-Boc-Propargylamine | 92136-39-5 | >99.78% | C8H13NO2 | 50 G
SDP
Catalog No. 5000434333
Encompass_Preferred
N-Boc-Propargylamine is a biochemical reagent used in life science research. It functions as a click chemistry reagent due to its alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Can be used as a biological material or organic compound
  • Contains an Alkyne group for click chemistry reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used to prepare triazolobenzylidene-thiazolopyrimidines (CDC25 phosphatase inhibitors)
  • Synthesizes beta-glucan polysaccharide analogs
  • Involved in Pauson-Khand reaction to prepare 4,5-dissubstituted cyclopentenones
Catalog No. 50-004-34333 Supplier Medchemexpress LLC Supplier No. HY7597050G
N-Boc-Propargylamine is a biochemical reagent used in life science research. It functions as a click chemistry reagent due to its alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Can be used as a biological material or organic compound
  • Contains an Alkyne group for click chemistry reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used to prepare triazolobenzylidene-thiazolopyrimidines (CDC25 phosphatase inhibitors)
  • Synthesizes beta-glucan polysaccharide analogs
  • Involved in Pauson-Khand reaction to prepare 4,5-dissubstituted cyclopentenones