Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-pyrrole

Catalog No. 42583425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-583-425ML 25 mL
42-583-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-583-425ML Supplier Sigma Aldrich Supplier No. 42583425ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-boc-pyrrole is an n-protected pyrrole. it undergoes diels-alder reaction with enantiomerically pure allene-1,3-dicarboxylates to form endo-adducts with retention in configurations at two newly generated stereogenic centers. it also undergoes cyclopropanation with methyl phenyldiazoacetate to form both monocyclopropane and dicyclopropane. its IR-catalyzed c-h borylation followed by cross coupling with 3-chlorothiophene to form biheterocycle has been reported.n-boc-pyrrole was used in the synthesis of 1-(tert-butoxycarbonyl)-1h-pyrrol-2-ylboronic acid by treating with n-buli and subsequent reaction with trimethyl borate.it may be used as starting material in the synthesis of the following:
  • Tropane drivatives
  • N-boc-2-(4-methoxyphenyl)pyrrole
  • N-boc-pyrrol-2-ylboronic acid

Specifications

CAS 5176-27-2
Density 1 g/mL (at 25°C (literature))
Boiling Point 91°C to 92°C (20 mmHg)
Molecular Formula C9H13NO2
Linear Formula C9H13NO2
Refractive Index n20/D 1.4685 (literature)
MDL Number MFCD00209559
Quantity 25 mL
Synonym tert-Butyl 1-pyrrolecarboxylate
Molecular Weight (g/mol) 167.21
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.