Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-S-ethyl-L-cysteine

Catalog No. AAH5021603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5021603 1 g
AAH5021606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5021603 Supplier Thermo Scientific Chemicals Supplier No. H5021603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 16947-82-3 | C10H19NO4S | 249.325 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16947-82-3
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.325
MDL Number MFCD00076919
InChI Key IBCCMMVPGKVLAX-ZETCQYMHSA-N
Synonym boc-cys et-oh, boc-s-ethyl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-ethylthio propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-ethylsulfanyl propanoic acid, n-boc-s-ethyl-l-cysteine, s-ethyl-n-t-butoxycarbonyl-l-cysteine, s-ethyl-n-t-butyloxycarbonyl-l-cysteine, n-tert-butoxycarbonyl-s-ethyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-ethyl
PubChem CID 7018740
IUPAC Name (2R)-3-ethylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CCSCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 249.33
Chemical Name or Material N-Boc-S-ethyl-L-cysteine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.