Learn More
Medchemexpress LLC N-Boc-trans-4-fluoro-L-proline | 203866-14-2 | MFCD06796085 | 233.24 g/mol | C10H16FNO4 | 10 G
SDP
Catalog No. 5000210223
Encompass_Preferred
N-Boc-trans-4-fluoro-L-proline is a Boc-protected, non-natural proline derivative bearing a 4-fluoro substituent on the pyrrolidine ring. It is used as a building block in peptide synthesis and for incorporation into peptides or protein constructs to modify conformation and physicochemical properties.
  • Boc-protected proline derivative for peptide synthesis.
  • Introduces a 4-fluoro substituent at the proline 4-position.
  • Suitable for incorporation into peptides and protein constructs.
  • Supplied in laboratory pack sizes, including 10 G, for research use.
  • Provided with standard identifiers (CAS and MFCD) for unambiguous identification.
Catalog No. 50-002-10223 Supplier Medchemexpress LLC Supplier No. HYW04607110G
N-Boc-trans-4-fluoro-L-proline is a Boc-protected, non-natural proline derivative bearing a 4-fluoro substituent on the pyrrolidine ring. It is used as a building block in peptide synthesis and for incorporation into peptides or protein constructs to modify conformation and physicochemical properties.
  • Boc-protected proline derivative for peptide synthesis.
  • Introduces a 4-fluoro substituent at the proline 4-position.
  • Suitable for incorporation into peptides and protein constructs.
  • Supplied in laboratory pack sizes, including 10 G, for research use.
  • Provided with standard identifiers (CAS and MFCD) for unambiguous identification.