Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-boc-trans-4-fluoro-L-proline | 203866-14-2 | MFCD06796085 | 98.0% | 233.24 g/mol | C10H16FNO4 | 1 ML
SDP

Catalog No. 5000210190
Encompass_Preferred

N-Boc-trans-4-fluoro-L-proline is a Boc-protected, non-natural L-proline derivative used as an amino-acid substitute in peptide synthesis and antibody engineering. It is supplied as a white to off-white solid or as a 10 mM solution in DMSO and is intended for research use only.

  • Boc-protected 4-fluoro L-proline derivative suitable for peptide synthesis and incorporation into peptides.
  • Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • Purity ≥98.0% (manufacturer); alternative assays report 97%.
  • Molecular weight 233.24 g/mol; formula C10H16FNO4.
  • Highly soluble in DMSO; sonication may be required for complete dissolution.
  • Store powder at -20°C (long term) or 4°C; store solutions at -80°C or -20°C per SDS recommendations.
  • For research use only; follow SDS for handling, storage, and disposal.

Catalog No. 50-002-10190 Supplier Medchemexpress LLC Supplier No. HYW04607110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-trans-4-fluoro-L-proline is a Boc-protected, non-natural L-proline derivative used as an amino-acid substitute in peptide synthesis and antibody engineering. It is supplied as a white to off-white solid or as a 10 mM solution in DMSO and is intended for research use only.

  • Boc-protected 4-fluoro L-proline derivative suitable for peptide synthesis and incorporation into peptides.
  • Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • Purity ≥98.0% (manufacturer); alternative assays report 97%.
  • Molecular weight 233.24 g/mol; formula C10H16FNO4.
  • Highly soluble in DMSO; sonication may be required for complete dissolution.
  • Store powder at -20°C (long term) or 4°C; store solutions at -80°C or -20°C per SDS recommendations.
  • For research use only; follow SDS for handling, storage, and disposal.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.