Learn More
Medchemexpress LLC N-Boc-Tris | 146651-71-0 | ≥97.0% | C9H19NO5 | 5 G
SDP
Catalog No. 5000355715
Encompass_Preferred
N-Boc-Tris is a protective reagent used to protect amino groups in organic synthesis and pharmaceutical research and development, preventing unwanted side reactions. It utilizes the Boc protecting group to safeguard amino groups in various chemical processes.
  • Protective reagent for amino groups
  • Prevents unwanted side reactions
  • Used in organic synthesis and pharmaceutical research
  • Appears as a white to off-white solid
  • Molecular weight: 221.25
Catalog No. 50-003-55715 Supplier Medchemexpress LLC Supplier No. HY1412535G
N-Boc-Tris is a protective reagent used to protect amino groups in organic synthesis and pharmaceutical research and development, preventing unwanted side reactions. It utilizes the Boc protecting group to safeguard amino groups in various chemical processes.
  • Protective reagent for amino groups
  • Prevents unwanted side reactions
  • Used in organic synthesis and pharmaceutical research
  • Appears as a white to off-white solid
  • Molecular weight: 221.25