Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Bromosuccinimide, 99%

Catalog No. ac107450100
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107450100 10 g Glass bottle
AC107451000 100 g Glass bottle
AC107455000 500 g Glass bottle
AC107450010 1 kg Glass bottle
AC107450050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. AC107450100 Supplier Thermo Scientific Chemicals Supplier No. 107450100
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent, C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-08-5
MDL Number MFCD00005510
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
Synonym n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
PubChem CID 67184
ChEBI CHEBI:53174
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 175.0°C to 178.0°C
Color White to Yellow
Flash Point 180°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Iodometry)
Packaging Glass bottle
Molecular Formula C4H4BrNO2
Quantity 10 g
Beilstein 21, 380
Fieser 01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49
Merck Index 15, 1448
Solubility Information Solubility in water: 14.8g/L (20°C). Other solubilities: 3.10g/100g glacial acetic acid(25°C),g/100g solvent (25°C):,tert. butanol:0.73- aceton:14.40,carbone tetrachloride:0.02- hexane:0.006,benzene:0.5
Molecular Weight (g/mol) 177.99
Formula Weight 177.99
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Bromosuccinimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire rem
EINECSNumber 204-877-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.