Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butanol, 7.5% v/v, Fisher Chemical™
SDP

Catalog No. SB95751 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB95751 1 L Amber Glass Bottle
SB9575-4 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB95751
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-36-3 , 7732-18-5 , 497-19-8 , 144-55-8
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 92.0, 5.54, 8.0E-4, 7.0E-4
CAS Max % 96.0, 5.77, 8.0E-4, 7.0E-4
Quantity 1 L
Packaging Amber Glass Bottle
Chemical Name or Material n-Butanol, 7.5% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.