Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butanol, 99% max., Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19400191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400191 1 L
ICN19400125 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19400191 Supplier MP Biomedicals, Inc Supplier No. 0219400191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Density 0.81g/cm3 at 21°C
Quantity 1 L
Physical Form Liquid
Formula Weight 74.12
Percent Purity 99% max.
Grade Molecular Biology
Chemical Name or Material n-Butanol
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.