Learn More
Medchemexpress LLC N-Butanoyl-DL-homoserine lactone | 98426-48-3 | 98.0% | 171.19 | 25 MG
SDP
Catalog No. 5000437636
Encompass_Preferred
N-Butanoyl-DL-homoserine lactone ((Rac)-C4-HSL) is a racemic mixture of N-Butanoyl-D-homoserine lactone and N-Butanoyl-L-homoserine lactone. This compound acts as a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) and demonstrates antibacterial activity, making it suitable for antibacterial biofilm applications.
  • Antibacterial activity
  • Used in antibacterial biofilm applications
  • Blocks quorum sensing
  • Inhibits biofilm formation in *Pseudomonas aeruginosa*
Catalog No. 50-004-37636 Supplier Medchemexpress LLC Supplier No. 25MGHY113764
N-Butanoyl-DL-homoserine lactone ((Rac)-C4-HSL) is a racemic mixture of N-Butanoyl-D-homoserine lactone and N-Butanoyl-L-homoserine lactone. This compound acts as a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) and demonstrates antibacterial activity, making it suitable for antibacterial biofilm applications.
  • Antibacterial activity
  • Used in antibacterial biofilm applications
  • Blocks quorum sensing
  • Inhibits biofilm formation in *Pseudomonas aeruginosa*