Learn More
Medchemexpress LLC N-butyl-3-[(4-decoxybenzoyl)amino]-4-methylsulfanylbenzamide | 135239-65-5 | MFCD00902351 | 99.6% | 498.72 g/mol | C29H42N2O3S | 10 MG
SDP
Catalog No. 5000330266
Encompass_Preferred
RP-64477 is a potent small-molecule inhibitor of Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT) provided for research use. It is supplied as a white to off-white solid with characterized physicochemical data suitable for biochemical and cellular studies.
  • Potent ACAT inhibition for cholesterol metabolism studies.
  • High purity (99.62%) suitable for research applications.
  • Supplied as a white to off-white solid with known molecular formula and weight.
  • Soluble in DMSO at 8.33 mg/mL to facilitate assay preparation.
  • Stable under recommended storage conditions to preserve compound integrity.
Catalog No. 50-003-30266 Supplier Medchemexpress LLC Supplier No. HY1643710MG
RP-64477 is a potent small-molecule inhibitor of Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT) provided for research use. It is supplied as a white to off-white solid with characterized physicochemical data suitable for biochemical and cellular studies.
  • Potent ACAT inhibition for cholesterol metabolism studies.
  • High purity (99.62%) suitable for research applications.
  • Supplied as a white to off-white solid with known molecular formula and weight.
  • Soluble in DMSO at 8.33 mg/mL to facilitate assay preparation.
  • Stable under recommended storage conditions to preserve compound integrity.