Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl acetate, 99+%

Catalog No. AAA194120F
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA194120F 2500 mL
AAA19412AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA194120F Supplier Thermo Scientific Chemicals Supplier No. A194120F
  • $172.20
    $145.65 / Each of 1

    Save $26.55

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In pharmaceutical and petroleum industries, it is used for extraction and as a processing solvent. It is an ingredient of cleaners, essences and fragrances. It is used as an analytical reagent to verify thalium, stannum and tungsten as well as in the determination of molybdenum and rhenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In pharmaceutical and petroleum industries, it is used for extraction and as a processing solvent. It is an ingredient of cleaners, essences and fragrances. It is used as an analytical reagent to verify thalium, stannum and tungsten as well as in the determination of molybdenum and rhenium.

Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone, chloroform and most hydrocarbons. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Density 0.881
pH 5
Boiling Point 124°C to 126°C
Flash Point 25°C (77°F)
Odor Fruit-like
Refractive Index 1.3945
Linear Formula CH3CO2(CH2)3CH3
Quantity 2500 mL
UN Number UN1123
Beilstein 1741921
Merck Index 14,1535
Solubility Information Miscible with ethanol,ethyl ether,alcohol,acetone,chloroform and most hydrocarbons. Slightly miscible with water.
Formula Weight 116.16
Percent Purity ≥99%
Chemical Name or Material n-Butyl acetate
Show More Show Less
Hazard Category H226-H336
Hazard Statement GHS H Statement
H226-H336
Flammable liquid and vapour.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES
EINECSNumber 204-658-1
RTECSNumber AF7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.