Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ n-Butyl acetate, ACS Grade, 99.5+%

Catalog No. AA46186AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46186AP 500 mL
AA46186K2 1 L
AA46186K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46186AP Supplier Thermo Scientific™ Supplier No. 46186AP
Add to Cart

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°
Density 0.881
Boiling Point 124-126°
Flash Point 25°(77°F)
Linear Formula CH{3}CO{2}(CH{2}){3}CH{3}
Quantity 500 mL
UN Number UN1123
Merck Index 14,1535
Formula Weight 116.16
Percent Purity 99.5+%
Physical Form Liquid
Show More Show Less
ShelfLife 3 Years
EINECSNumber 204-658-1
RTECSNumber AF350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.