Learn More
Electron Microscopy Sciences N-Butyl Alcohol 450 ML
SDP
Catalog No. 5030132
Encompass_Preferred
Phloxine B Staining Solution is used in relative aciddophilia stain. The least acidophilic elements stained in the phloxine, control slide lose the red color after 2 minutes in the tartrazine solution. The most acidophilic retain the phloxine after 20 minutes in the tartrazine.
References: Lendrum, A.C, J. Path. Bact., 59:399-404, 1947 and Clark, G., (ed.), Staining Procedures, 3rd Edition, Williams & Wilkins, Baltimore, p. 163, c. 1973.
Catalog No. 50-301-32 Supplier Electron Microscopy Sciences Supplier No. 2690103
Phloxine B Staining Solution is used in relative aciddophilia stain. The least acidophilic elements stained in the phloxine, control slide lose the red color after 2 minutes in the tartrazine solution. The most acidophilic retain the phloxine after 20 minutes in the tartrazine.
References: Lendrum, A.C, J. Path. Bact., 59:399-404, 1947 and Clark, G., (ed.), Staining Procedures, 3rd Edition, Williams & Wilkins, Baltimore, p. 163, c. 1973.