Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Butyl Alcohol, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000550 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000550 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: Flammable liquid and vapour, harmful if swallowed, causes skin irritation, causes serious eye damage, may cause respiratory irritation, may cause drowsiness or dizziness.
Catalog No. S000550 Supplier Aldon Corporation Supplier No. IS11077
Add to Cart
Add to Cart
Danger: Flammable liquid and vapour, harmful if swallowed, causes skin irritation, causes serious eye damage, may cause respiratory irritation, may cause drowsiness or dizziness.

Chemical Identifiers

Molecular Weight (g/mol) 74.12 g/mol

Specifications

Boiling Point 117.5°C
Quantity 500 mL
Percent Purity > 99.8
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.