Learn More
Medchemexpress LLC n-Butyl benzoate | 136-60-7 | MFCD00009439 | 99.7% | 178.23 g·mol⁻¹ | C11H14O2 | 1 KG
SDP
Catalog No. 5000301149
Encompass_Preferred
Butyl benzoate (n-butyl benzoate) is an aromatic ester used as a solvent, fragrance ingredient, and chemical intermediate. It is supplied as a clear, high-purity liquid suitable for research, formulation, and analytical applications.
  • High purity (99.73%) liquid for research and formulation use.
  • Used as a solvent and perfume ingredient in fragrance and flavor applications.
  • Low volatility and good solvency for aromatic compounds.
  • Soluble in common organic solvents and DMSO for analytical preparations.
  • Available in multiple pack sizes for lab-scale use.
Catalog No. 50-003-01149 Supplier Medchemexpress LLC Supplier No. HYW0165991KG
Butyl benzoate (n-butyl benzoate) is an aromatic ester used as a solvent, fragrance ingredient, and chemical intermediate. It is supplied as a clear, high-purity liquid suitable for research, formulation, and analytical applications.
  • High purity (99.73%) liquid for research and formulation use.
  • Used as a solvent and perfume ingredient in fragrance and flavor applications.
  • Low volatility and good solvency for aromatic compounds.
  • Soluble in common organic solvents and DMSO for analytical preparations.
  • Available in multiple pack sizes for lab-scale use.