Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Chloride, for HPLC, GC and spectrophotometry, ≥99.5%, Solstice

Catalog No. 6000097 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-000-97 6 x 1 L Glass Bottle
60-000-96 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-000-97 Supplier Solstice Supplier No. 0341L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

CAS Max % 100.0000%
Density 0.8862 g/cm3
Boiling Point 78°C
Flash Point −9°C
Loss on Drying 1 mg/L max.
Packaging Glass Bottle
Quantity 6 x 1 L
UN Number UN1127
Beilstein 1730909
Solubility Information 11% in Water at 20°C
Formula Weight 92.58
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material n-Butyl Chloride, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Chlorobutanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.