Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-butyl cinnamate | 538-65-8 | MFCD00051560 | 99.7% | 204.27 g·mol⁻1 | C13H16O2 | 10MM 1ML
SDP

Catalog No. 5000236095
Encompass_Preferred

N-Butyl cinnamate (CAS 538-65-8) is an organic ester provided as a ready-to-use 10 mM solution in DMSO for laboratory research. It is intended for biochemical and cellular assays and has reported antiinflammatory activity in endothelial cell models.

  • Ready-to-use 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.65%) suitable for research applications.
  • Molecular formula C13H16O2; molecular weight 204.27 g·mol⁻1.
  • Supplied with data sheet, SDS, COA, HNMR, and GC files.
  • Storage recommendations provided for pure material and solvent solutions.
  • Reported antiinflammatory activity in HUVEC assays (IC50 125 μg/mL).

Catalog No. 50-002-36095 Supplier Medchemexpress LLC Supplier No. HY12607010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Butyl cinnamate (CAS 538-65-8) is an organic ester provided as a ready-to-use 10 mM solution in DMSO for laboratory research. It is intended for biochemical and cellular assays and has reported antiinflammatory activity in endothelial cell models.

  • Ready-to-use 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.65%) suitable for research applications.
  • Molecular formula C13H16O2; molecular weight 204.27 g·mol⁻1.
  • Supplied with data sheet, SDS, COA, HNMR, and GC files.
  • Storage recommendations provided for pure material and solvent solutions.
  • Reported antiinflammatory activity in HUVEC assays (IC50 125 μg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.