Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Glycidyl Ether, MP Biomedicals

Catalog No. ICN211771 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211771 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211771 Supplier MP Biomedicals, Inc Supplier No. 0521177180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2426-08-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
InChI Key YSUQLAYJZDEMOT-UHFFFAOYNA-N
Synonym butyl glycidyl ether, n-butyl glycidyl ether, 2-butoxymethyl oxirane, oxirane, butoxymethyl, 1-butoxy-2,3-epoxypropane, glycidyl butyl ether, butoxymethyl oxirane, ether, butyl glycidyl, glycidyl n-butyl ether, butyl 2,3-epoxypropyl ether
PubChem CID 17049
IUPAC Name 2-(butoxymethyl)oxirane
SMILES CCCCOCC1CO1

Specifications

Quantity 100 g
Formula Weight 130.1
Chemical Name or Material n-Butyl Glycidyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.