Learn More
Cayman Chemical N-butyl LIthocholIc AcId 10mg
SDP
Catalog No. 502944360
Encompass_Preferred
A derivative of lithocholic acid; LNPs containing N-butyl lithocholic acid and encapsulating an mRNA vaccine expressing influenza hemagglutinin exhibit less heart, spleen, and liver toxicity than LNPs containing cholesterol in mice
Catalog No. 50-294-4360 Supplier Cayman Chemical Supplier No. 4139710 MG
A derivative of lithocholic acid; LNPs containing N-butyl lithocholic acid and encapsulating an mRNA vaccine expressing influenza hemagglutinin exhibit less heart, spleen, and liver toxicity than LNPs containing cholesterol in mice