Learn More
Electron Microscopy Sciences N-Butyl Methacrylate 225 ML
SDP
Catalog No. 50980274
Encompass_Preferred
CH₂C(CH₃)CO₂CH₂CH₂CH₂CH₃ F.W. 142.20
Boiling point: 162-164°C
Stabilized with 10ppm MEHQ (Hydroquinone, Mononethylether).
Catalog No. 50-980-274 Supplier Electron Microscopy Sciences Supplier No. 12100
CH₂C(CH₃)CO₂CH₂CH₂CH₂CH₃ F.W. 142.20
Boiling point: 162-164°C
Stabilized with 10ppm MEHQ (Hydroquinone, Mononethylether).