Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Butyl-N-(4-hydroxybutyl)nitrosamine 90.0+%, TCI America™
SDP

Catalog No. B093825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B093825G 25 g
B09385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B093825G Supplier TCI America Supplier No. B093825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3817-11-6
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD00059008
InChI Key DIKPQFXYECAYPC-UHFFFAOYSA-N
Synonym N-Butyl-N-butan-4-ol-nitrosamine, N-Butyl-N-nitroso-4-aminobutanol, BBN
PubChem CID 19665
IUPAC Name N-butyl-N-(4-hydroxybutyl)nitrous amide
SMILES CCCCN(CCCCO)N=O

Specifications

Boiling Point 167°C
Quantity 25 g
Formula Weight 174.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Butyl-N-(4-hydroxybutyl)nitrosamine
RTECSNumber EL1225000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.