Learn More
Selleck Chemical LLC N-butyl-N- nitrosamine
SDP
Catalog No. 503137067
Encompass_Preferred
N-butyl-N- nitrosamine (BBN OH-BBN) is a model compound that induces high-grade invasive tumors in the urinary bladder N-butyl-N- nitrosamine can be used to induce animal models of Bladder Carcinoma
Catalog No. 50-313-7067 Supplier Selleck Chemical LLC Supplier No. E01991G
N-butyl-N- nitrosamine (BBN OH-BBN) is a model compound that induces high-grade invasive tumors in the urinary bladder N-butyl-N- nitrosamine can be used to induce animal models of Bladder Carcinoma