Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl propionate, 99+%

Catalog No. ac269482500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269482500 250 mL Glass bottle
AC269480010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC269482500 Supplier Thermo Scientific Chemicals Supplier No. 269482500
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 590-01-2 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -75°C
Color Colorless
Density 0.8800g/mL
Boiling Point 145°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4000 to 1.402
Linear Formula CH3(CH2)3OC(O)C2H5
Quantity 250 mL
Beilstein 02, 241
Specific Gravity 0.88
Merck Index 15, 1590
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.83 mPa.s (20°C)
Formula Weight 130.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butyl propionate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-669-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.