Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl sulfide, 99%, pure

Catalog No. AC108010025
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108010025 2.5 L Glass bottle
AC108011000 100 mL Glass bottle
AC108015000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108010025 Supplier Thermo Scientific Chemicals Supplier No. 108010025
  • $1,246.00
    $1,059.65 / Each of 1

    Save $186.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 544-40-1 | C8H18S | 146.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-40-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
Synonym dibutyl sulfide, butyl sulfide, n-butyl sulfide, butylthiobutane, di-n-butyl sulfide, butane, 1,1'-thiobis, butyl monosulfide, dibutyl thioether, thiononane-5, n-dibutyl sulfide
PubChem CID 11002
SMILES CCCCSCCCC

Specifications

Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Linear Formula [CH3(CH2)3]2S
Quantity 2.5 L
Beilstein 01,IV,1559
Merck Index 15,1593
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether,soluble in most organic solvents
IUPAC Name 1-butylsulfanylbutane
Formula Weight 146.29
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Butyl sulfide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-870-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.