Learn More
Cayman Chemical N-Butylfluorescein, Purity: ≥98%, CAS: 335193-91-4, 10 mg
SDP
Catalog No. 502984085
Encompass_Preferred
Ex/em = 467/512 nm, respectively; has been used in the synthesis of butyl FLIP, a fluorogenic substrate of PI-PLC
Catalog No. 50-298-4085 Supplier Cayman Chemical Supplier No. 2090910 MG
Ex/em = 467/512 nm, respectively; has been used in the synthesis of butyl FLIP, a fluorogenic substrate of PI-PLC