Learn More
Selleck Chemical LLC n-Butylidenephthalide
SDP
Catalog No. 5000003639
Encompass_Preferred
n-Butylidenephthalide ((Z)-Butylidenephthalide 3-Butylidenephthalide Butylidenephthalide) isolated from the chloroform extract of Angelica sinensis (AS-C) contributes to a range of biological activities including diminution of angina platelet aggregation proliferation non-specific spasmosis
Catalog No. 50-000-03639 Supplier Selleck Chemical LLC Supplier No. S649325MG
n-Butylidenephthalide ((Z)-Butylidenephthalide 3-Butylidenephthalide Butylidenephthalide) isolated from the chloroform extract of Angelica sinensis (AS-C) contributes to a range of biological activities including diminution of angina platelet aggregation proliferation non-specific spasmosis