Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyllithium, 2.2M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AA412483Y
Change view
Click to view available options
Quantity:
0.5 mol
Packaging:
Packaged under Argon in resealable ChemSeal™ bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA412483Y 0.5 mol Packaged under Argon in resealable ChemSeal™ bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA412483Y Supplier Thermo Scientific Chemicals Supplier No. 412483Y

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-72-8
Flash Point −21°C (−7°F)
Odor Petroleum-like
Packaging Packaged under Argon in resealable ChemSeal™ bottles
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
Quantity 0.5 mol
MDL Number MFCD00009414
UN Number UN3394
Beilstein 1209227
Sensitivity Air and moisture sensitive
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
Solubility Information Miscible with diethyl ether and cyclohexane.
InChI Key DLEDOFVPSDKWEF-UHFFFAOYSA-N
SMILES [Li+].CCC[CH2-]
IUPAC Name lithium;butane
Molecular Weight (g/mol) 64.056
PubChem CID 61028
Formula Weight 64.06
Chemical Name or Material n-Butyllithium
Show More Show Less
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336;
Highly flammable liquid and vapour.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 203-698-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.