Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Butyllithium Solution

Catalog No. 503081484 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
8 L
18 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1484 4 x 25 mL
23-070-78L 8 L
50-308-1482 100 mL
50-308-1483 4 x 100 mL
23-070-718LKL 18 L
23-070-7800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Danger
Catalog No. 50-308-1484 Supplier Sigma Aldrich Supplier No. 2307074X25ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 109-72-8
Density 0.693 g/mL at 25°C
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
Quantity 4 x 25 mL
Synonym n-BuLi; Butyl lithium; Butyllithium solution; Lithium-1-butanide
Molecular Weight (g/mol) 64.06 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.