Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyllithium, Thermo Scientific™

Catalog No. AC213351300
Catalog No. AC213351300 Supplier Thermo Scientific Chemicals Supplier No. 213351300

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-72-8 , 110-54-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
IUPAC Name butyllithium
SMILES [Li]CCCC

Specifications

Boiling Point 60.0°C to 80.0°C
Melting Point -95.0°C
Linear Formula CH3(CH2)3Li
Density 0.6900g/mL
Solubility Information Solubility in water: reacts with water forming lioh and butane.
Formula Weight 64.06
Percent Purity 22.0 to 24.0% (Active base) 2.37 to 2.59 M (Active base)
Specific Gravity 0.69
Flash Point −21°C
Chemical Name or Material n-Butyllithium
Hazard Category Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. May cause drowsiness or dizziness. Suspected of damaging fertility. Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.