Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Butyllithium, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 60.0°C to 80.0°C |
| Melting Point | -95.0°C |
| Linear Formula | CH3(CH2)3Li |
| Density | 0.6800g/mL |
| Solubility Information | Solubility in water: reacts (forming lioh an butane). |
| Formula Weight | 64.06 |
| Percent Purity | 14.0 to 16.0% (Active base) 1.49 to 1.70 M (Active base) |
| Specific Gravity | 0.68 |
| Flash Point | −18°C |
| Chemical Name or Material | n-Butyllithium |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Reacts violently with water. |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction