Learn More
Medchemexpress LLC N-butylthiophosphoric triamide | 94317-64-3 | MFCD00269941 | 99.8% | 167.21 | C4H14N3PS | 10 G
SDP
Catalog No. 5000240432
Encompass_Preferred
N-Butylthiophosphoric triamide (NBPT) is a urease inhibitor used to delay urea hydrolysis and reduce ammonia volatilization from nitrogen fertilizers. It binds to the urease active site, forming a stable complex that prevents urea from accessing the catalytic nickel center, improving nitrogen-use efficiency and helping to reduce environmental nitrogen loss.
  • Potent urease inhibitor that reduces conversion of urea to ammonia.
  • Binds urease active site to delay urea hydrolysis.
  • Improves nitrogen-use efficiency and can increase crop yield.
  • Reduces ammonia volatilization and associated environmental loss.
  • High purity solid formulation (~99.8%).
  • Available as solid and solution formulations; store at 4°C and protect from light.
Catalog No. 50-002-40432 Supplier Medchemexpress LLC Supplier No. HY10719910G
N-Butylthiophosphoric triamide (NBPT) is a urease inhibitor used to delay urea hydrolysis and reduce ammonia volatilization from nitrogen fertilizers. It binds to the urease active site, forming a stable complex that prevents urea from accessing the catalytic nickel center, improving nitrogen-use efficiency and helping to reduce environmental nitrogen loss.
  • Potent urease inhibitor that reduces conversion of urea to ammonia.
  • Binds urease active site to delay urea hydrolysis.
  • Improves nitrogen-use efficiency and can increase crop yield.
  • Reduces ammonia volatilization and associated environmental loss.
  • High purity solid formulation (~99.8%).
  • Available as solid and solution formulations; store at 4°C and protect from light.