Learn More
Medchemexpress LLC N-butylthiophosphoric triamide | 94317-64-3 | MFCD00269941 | 99.8% | 167.22 g/mol | C4H14N3PS | 5 G
SDP
Catalog No. 5000240931
Encompass_Preferred
N-Butylthiophosphoric triamide is an organophosphorus urease inhibitor that slows enzymatic hydrolysis of urea, reducing ammonia volatilization and improving nitrogen retention. It is provided in solid form and as DMSO solutions for laboratory and formulation use.
  • Reduces urea hydrolysis and ammonia loss
  • Improves nitrogen use efficiency in agricultural applications
  • Available as solid and DMSO solution formats for flexibility
  • High purity suitable for analytical and research use
  • Safety data sheet and technical documentation available
Catalog No. 50-002-40931 Supplier Medchemexpress LLC Supplier No. HY1071995G
N-Butylthiophosphoric triamide is an organophosphorus urease inhibitor that slows enzymatic hydrolysis of urea, reducing ammonia volatilization and improving nitrogen retention. It is provided in solid form and as DMSO solutions for laboratory and formulation use.
  • Reduces urea hydrolysis and ammonia loss
  • Improves nitrogen use efficiency in agricultural applications
  • Available as solid and DMSO solution formats for flexibility
  • High purity suitable for analytical and research use
  • Safety data sheet and technical documentation available