Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyric Acid, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604766 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-766 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-766 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B23071KGBL

missing translation for 'validMainframeMsg'

B2307, 107-92-6, C4H8O2

Spectrum™ Chemical n-Butyric Acid, FCC is used as an animal feed supplement due to the ability to reduce pathogenic bacterial colonization and is an approved food flavoring in the EU FLAVIS database (number 08.005). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

CAS Max % 1
Quantity 1 kg
Packaging Amber Glass Bottle
Refractive Index 1.397 to 1.399
Specific Gravity 0.953-0.957
Percent Purity 99%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.