Learn More
AB Sciex LLC N/C FUSED SILICA CAPILLARY 3/P
Catalog No. NC9812569
Encompass
Untreated Fused-Silica Capillaries 50cm effective length 50micrometer I.D 375micrometer O.D
Catalog No. NC9812569 Supplier AB Sciex LLC Supplier No. 338451
Untreated Fused-Silica Capillaries 50cm effective length 50micrometer I.D 375micrometer O.D