Learn More
Medchemexpress LLC N-Cadherin Human H 10ug
SDP
Catalog No. 5000181222
Encompass_Preferred
N-Cadherin Protein forms dimers with CDH2 chains promoting cell-cell adhesion and sorting of cell types It regulates neural stem cell quiescence by anchoring them to ependymocytes disrupted by MMP24 cleavage It also promotes cell-to-cell junctions and neurite branching and regulates dendritic spine density N-Cadherin Protein interacts with CDCP1 PCDH8 OBSCN FGFR4 NCAM1 CDH2 PLCG1 FRS2 SRC SHC1 GAP43 CTTN and FBXO45 N-Cadherin Protein Human (699a a HEK293 His) N-Cadherin Protein Human (HEK293 His) is the recombinant human-derived N-Cadherin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81222 Supplier Medchemexpress LLC Supplier No. HYP7330210UG
N-Cadherin Protein forms dimers with CDH2 chains promoting cell-cell adhesion and sorting of cell types It regulates neural stem cell quiescence by anchoring them to ependymocytes disrupted by MMP24 cleavage It also promotes cell-to-cell junctions and neurite branching and regulates dendritic spine density N-Cadherin Protein interacts with CDCP1 PCDH8 OBSCN FGFR4 NCAM1 CDH2 PLCG1 FRS2 SRC SHC1 GAP43 CTTN and FBXO45 N-Cadherin Protein Human (699a a HEK293 His) N-Cadherin Protein Human (HEK293 His) is the recombinant human-derived N-Cadherin protein expressed by HEK293 with C-His labeled tag