Learn More
Medchemexpress LLC N-Cadherin Mouse H 10ug
SDP
Catalog No. 5000179563
Encompass_Preferred
N-Cadherin Protein is a calcium-dependent cell adhesion protein that mediates homotypic cell-cell adhesion by dimerizing with a CDH2 chain from another cell N-Cadherin Protein plays a role in cell adhesion cell sorting and neural development N-Cadherin Protein Mouse (HEK293) is the recombinant mouse-derived N-Cadherin protein expressed by HEK293 with tag free
Catalog No. 50-001-79563 Supplier Medchemexpress LLC Supplier No. HYP7375010UG
N-Cadherin Protein is a calcium-dependent cell adhesion protein that mediates homotypic cell-cell adhesion by dimerizing with a CDH2 chain from another cell N-Cadherin Protein plays a role in cell adhesion cell sorting and neural development N-Cadherin Protein Mouse (HEK293) is the recombinant mouse-derived N-Cadherin protein expressed by HEK293 with tag free