Learn More
Enzo Life Sciences N-cadherin polyclonal antibody (400µg)
SDP
Supplier:  Enzo Life Sciences ALX210618C400
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 808-827 of human N-cadherin. UniProt ID: P19022. Species reactivity: Human, Mouse Chicken, Bovine. Creossreactivity: Does not cross-react with human E- or P-cadherin. Applications: IHC (FS), IHC (PS), WB. Recommended Dilutions: Immunohistochemistry (frozen and paraffin sections, 5-10µg/ml) Western Blot (5-10µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Lyophilized from 10mM PBS, pH 7.4,  containing 1% BSA. Does not contain preservatives. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8136