Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 1 ML
SDP

Catalog No. 5000419107
Encompass_Preferred

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa that demonstrates strong inhibitory activity against α-glucosidase. It has a molecular weight of 313.35 and a chemical formula of C18H19NO4. This product is supplied as a 10 mM solution in 1 mL for research use only.

  • Strong inhibitory activity against α-glucosidase
  • Molecular weight of 313.35
  • Chemical formula C18H19NO4
  • Isolated from Cuscuta reflexa
  • For research use only
  • Supplied as a 10 mM solution in 1 mL

Catalog No. 50-004-19107 Supplier Medchemexpress LLC Supplier No. HYN720310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa that demonstrates strong inhibitory activity against α-glucosidase. It has a molecular weight of 313.35 and a chemical formula of C18H19NO4. This product is supplied as a 10 mM solution in 1 mL for research use only.

  • Strong inhibitory activity against α-glucosidase
  • Molecular weight of 313.35
  • Chemical formula C18H19NO4
  • Isolated from Cuscuta reflexa
  • For research use only
  • Supplied as a 10 mM solution in 1 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.