Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 10 MG
SDP

Catalog No. 5000417698
Encompass_Preferred

N-Caffeoyl O-methyltyramine is an alkaloid isolated from *Cuscuta reflexa* that demonstrates strong inhibitory activity against α-glucosidase. This product is for research use only.

  • Inhibits α-glucosidase (IC50: 103.58 μM)
  • Molecular weight: 313.35
  • Molecular formula: C18H19NO4
  • Appearance: White to off-white solid
  • Purity: 98.52%
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-17698 Supplier Medchemexpress LLC Supplier No. HYN720310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is an alkaloid isolated from *Cuscuta reflexa* that demonstrates strong inhibitory activity against α-glucosidase. This product is for research use only.

  • Inhibits α-glucosidase (IC50: 103.58 μM)
  • Molecular weight: 313.35
  • Molecular formula: C18H19NO4
  • Appearance: White to off-white solid
  • Purity: 98.52%
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.