Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Caffeoyl O-methyltyramine | 189307-47-9 | 100 MG
SDP

Catalog No. 5000418901
Encompass_Preferred

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa, known for its strong inhibitory activity against α-glucosidase (IC50 of 103.58 μM). It is supplied as a solid with a purity of 98.52% and a molecular weight of 313.35. The compound's formula is C18H19NO4.

  • Alkaloid isolated from *Cuscuta reflexa*
  • Strong α-glucosidase inhibitory activity
  • Purity of 98.52%
  • Molecular formula C18H19NO4
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-18901 Supplier Medchemexpress LLC Supplier No. HYN7203100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Caffeoyl O-methyltyramine is an alkaloid isolated from Cuscuta reflexa, known for its strong inhibitory activity against α-glucosidase (IC50 of 103.58 μM). It is supplied as a solid with a purity of 98.52% and a molecular weight of 313.35. The compound's formula is C18H19NO4.

  • Alkaloid isolated from *Cuscuta reflexa*
  • Strong α-glucosidase inhibitory activity
  • Purity of 98.52%
  • Molecular formula C18H19NO4
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.